AC5H93DE82FF Accela ChemBio Inc.

9-Bromo-2-iodo-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepine

  • MDL number MFCD22690733
  • Cas Number : 1282516-69-1
  • Molecular Weight : 391.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-bromo-2-iodo-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepine
Inchi Code 1S/C11H8BrIN2O/c12-7-1-2-8-9(5-7)16-4-3-15-6-10(13)14-11(8)15/h1-2,5-6H,3-4H2
Inchi Key SMHDMAYAZJZZHS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY071363&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY071363.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart