AC5H93DE8302 Accela ChemBio Inc.

N,N-Diethyl-4-nitrobenzamide

  • MDL number MFCD00454141
  • Cas Number : 5323-47-7
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N-diethyl-4-nitrobenzamide
Inchi Code 1S/C11H14N2O3/c1-3-12(4-2)11(14)9-5-7-10(8-6-9)13(15)16/h5-8H,3-4H2,1-2H3
Inchi Key JLZWKZDHHREXTA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY076075&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280a-P305+P351+P338-P304+P340-P405-P501a
MSDS http://pdf.accelachem.com/cn/msds/SY076075.pdf
hcodes GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart