AC5H93DE8304 Accela ChemBio Inc.

2-Amino-1-propylbenzimidazole

  • MDL number MFCD00964693
  • Cas Number : 57667-50-2
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-propyl-1H-benzimidazol-2-ylamine
Inchi Code 1S/C10H13N3/c1-2-7-13-9-6-4-3-5-8(9)12-10(13)11/h3-6H,2,7H2,1H3,(H2,11,12)
Inchi Key MGKUHRRJQCKXOW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY079922&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY079922.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart