AC5H93DE831F Accela ChemBio Inc.

2-Nitro-1-naphthalenamine

  • MDL number MFCD00021543
  • Cas Number : 607-23-8
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-nitro-1-naphthalenamine
Inchi Code 1S/C10H8N2O2/c11-10-8-4-2-1-3-7(8)5-6-9(10)12(13)14/h1-6H,11H2
Inchi Key SMAJHKXZBICXLS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129316&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129316.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart