AC5H93DE8321 Accela ChemBio Inc.

2-Amino-6-bromo-5-methyl-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD18642392
  • Cas Number : 1124382-65-5
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-5-methyl[1,2,4]triazolo[1,5-a]pyridin-2-amine
Inchi Code 1S/C7H7BrN4/c1-4-5(8)2-3-6-10-7(9)11-12(4)6/h2-3H,1H3,(H2,9,11)
Inchi Key ZNPGWTXQRAGCLF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129520&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129520.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart