AC5H93DE8327 Accela ChemBio Inc.

N6-Cbz-N2,N2-bis(carboxymethyl)-L-lysine

  • MDL number MFCD27977806
  • Cas Number : 113231-04-2
  • Molecular Weight : 396.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-2,2'-((5-(((benzyloxy)carbonyl)amino)-1-carboxypentyl)azanediyl)diacetic acid
Inchi Code 1S/C18H24N2O8/c21-15(22)10-20(11-16(23)24)14(17(25)26)8-4-5-9-19-18(27)28-12-13-6-2-1-3-7-13/h1-3,6-7,14H,4-5,8-12H2,(H,19,27)(H,21,22)(H,23,24)(H,25,26)/t14-/m0/s1
Inchi Key RKSASMXBVVMAAS-AWEZNQCLSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY131107&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY131107.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart