AC5H93DE8336 Accela ChemBio Inc.

Dihydroresveratrol

  • MDL number MFCD25973128
  • Cas Number : 58436-28-5
  • Molecular Weight : 230.25916

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232649&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232649.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart