AC5H93DE833B Accela ChemBio Inc.

(S)-1-Amino-3-methoxy-2-propanol

  • MDL number MFCD09260574
  • Cas Number : 1085543-96-9
  • Molecular Weight : 105.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-1-amino-3-methoxypropan-2-ol
Inchi Code 1S/C4H11NO2/c1-7-3-4(6)2-5/h4,6H,2-3,5H2,1H3/t4-/m0/s1
Inchi Key IDSWHVZEQHESIJ-BYPYZUCNSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY236677&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY236677.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart