AC5H93DE833C Accela ChemBio Inc.

3,5-Dimethyl-1-trityl-1H-pyrazole-4-boronic Acid Pinacol Ester

  • MDL number MFCD31699909
  • Cas Number : 1874148-66-9
  • Molecular Weight : 464.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole
Inchi Code 1S/C30H33BN2O2/c1-22-27(31-34-28(3,4)29(5,6)35-31)23(2)33(32-22)30(24-16-10-7-11-17-24,25-18-12-8-13-19-25)26-20-14-9-15-21-26/h7-21H,1-6H3
Inchi Key AXWJCSNCJUEQKV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237245&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237245.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart