AC5H93DE833F Accela ChemBio Inc.

4-Methoxyquinoline

  • MDL number MFCD00483884
  • Cas Number : 607-31-8
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxyquinoline
Inchi Code 1S/C10H9NO/c1-12-10-6-7-11-9-5-3-2-4-8(9)10/h2-7H,1H3
Inchi Key RWTCJCUERNMVEZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid, or colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY239034&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY239034.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart