AC5H93DE8340 Accela ChemBio Inc.

2-(3-Bromophenyl)acetaldehyde

  • MDL number MFCD02261727
  • Cas Number : 109347-40-2
  • Molecular Weight : 199.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to tan liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY239726&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY239726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart