AC5H93DE834B Accela ChemBio Inc.

(S)-N-[1-(Dimethylamino)-3-phenyl-2-propyl]acetamide

  • MDL number MFCD31977865
  • Cas Number : 2226371-98-6
  • Molecular Weight : 220.3107

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form White to yellow solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248776&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248776.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart