AC5H93DE8356 Accela ChemBio Inc.

3,4-Bis(phenylsulfonyl)-1,2,5-oxadiazole 2-Oxide

  • MDL number MFCD29042396
  • Cas Number : 66074-00-8
  • Molecular Weight : 366.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-bis(phenylsulfonyl)-1,2,5-oxadiazole 2-oxide
Inchi Code 1S/C14H10N2O6S2/c17-16-14(24(20,21)12-9-5-2-6-10-12)13(15-22-16)23(18,19)11-7-3-1-4-8-11/h1-10H
Inchi Key PLIHJANRRMFJCC-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253266&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253266.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart