AC5H93DE835B Accela ChemBio Inc.

2-[(4-Aminopentyl)(ethyl)amino]ethanol

  • MDL number MFCD03788185
  • Cas Number : 69559-11-1
  • Molecular Weight : 174.28378

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Yellow to brown liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY254057&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY254057.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart