AC5H93DE835D Accela ChemBio Inc.

(S)-2,2',2''-[10-[2-[4-[3-[[4-[[2-(2-Cyano-4,4-difluoro-1-pyrrolidinyl)-2-oxoethyl]carbamoyl]-6-quinolyl]oxy]propyl]-1-piperazinyl]-2-oxoethyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triyl]triacetic Acid

  • MDL number MFCD32067986
  • Cas Number : 2374782-02-0
  • Molecular Weight : 872.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY257808&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY257808.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart