AC5H93DE835F Accela ChemBio Inc.

Ethyl 4,5,6,7-Tetrahydro-1H-indole-2-carboxylate

  • MDL number MFCD14635837
  • Cas Number : 65880-18-4
  • Molecular Weight : 193.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY258170&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY258170.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart