AC5H93DE8361 Accela ChemBio Inc.

Dimethyl 2-(3-Bromophenyl)malonate

  • MDL number MFCD06204200
  • Cas Number : 773134-24-0
  • Molecular Weight : 287.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dimethyl 2-(3-bromophenyl)malonate
Inchi Code 1S/C11H11BrO4/c1-15-10(13)9(11(14)16-2)7-4-3-5-8(12)6-7/h3-6,9H,1-2H3
Inchi Key JXNAZEYUONMIGH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY260139&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY260139.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart