AC5H93DE8364 Accela ChemBio Inc.

1,4-Bis(6-nitro-3-pyridyl)piperazine

  • MDL number MFCD32202760
  • Cas Number : 2340184-77-0
  • Molecular Weight : 330.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,4-bis(6-nitropyridin-3-yl)piperazine
Inchi Code 1S/C14H14N6O4/c21-19(22)13-3-1-11(9-15-13)17-5-7-18(8-6-17)12-2-4-14(16-10-12)20(23)24/h1-4,9-10H,5-8H2
Inchi Key QCQAAJKHXNYIQM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262064&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262064.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart