AC5H93DE836D Accela ChemBio Inc.

2,5-Bis(3,5-difluorophenyl)pyridine

  • MDL number MFCD32642024
  • Cas Number : 1627313-24-9
  • Molecular Weight : 303.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-bis(3,5-difluorophenyl)pyridine

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY264908&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY264908.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart