AC5H93DE836E Accela ChemBio Inc.

N-[2-(5-Imidazolyl)ethyl]acrylamide

  • MDL number MFCD21183796
  • Cas Number : 10124-85-3
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[2-(1H-imidazol-5-yl)ethyl]acrylamide
Inchi Code 1S/C8H11N3O/c1-2-8(12)10-4-3-7-5-9-6-11-7/h2,5-6H,1,3-4H2,(H,9,11)(H,10,12)
Inchi Key PDNUJRVEYKRJFO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY264918&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY264918.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart