AC5H93DE8379 Accela ChemBio Inc.

2-Amino-5-bromo-6-methoxynicotinic Acid

  • MDL number MFCD32642050
  • Cas Number : 1935891-49-8
  • Molecular Weight : 247.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY265464&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY265464.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart