AC5H93DE837C Accela ChemBio Inc.

Ethyl 1,1-Difluorospiro[2.5]octane-6-carboxylate

  • MDL number MFCD31650074
  • Cas Number : 1447942-59-7
  • Molecular Weight : 218.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1,1-difluorospiro[2.5]octane-6-carboxylate
Inchi Code 1S/C11H16F2O2/c1-2-15-9(14)8-3-5-10(6-4-8)7-11(10,12)13/h8H,2-7H2,1H3
Inchi Key UNXWRRRPQUEKEQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY265534&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY265534.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart