AC5H93DE8381 Accela ChemBio Inc.

tert-Butyl 2-(1-Boc-azetidin-3-ylidene)acetate

  • MDL number MFCD32642068
  • Cas Number : 1803090-73-4
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(2-(tert-butoxy)-2-oxoethylidene)azetidine-1-carboxylate
Inchi Code 1S/C14H23NO4/c1-13(2,3)18-11(16)7-10-8-15(9-10)12(17)19-14(4,5)6/h7H,8-9H2,1-6H3
Inchi Key DWPOSKWNJOUAFR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid, or colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY265849&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY265849.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart