AC5H93DE838A Accela ChemBio Inc.

2-Butyl-4-methoxy-1H-imidazole

  • MDL number MFCD32662114
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-butyl-4-methoxy-1H-imidazole

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY266258&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY266258.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart