AC5H93DE8392 Accela ChemBio Inc.

(S)-3-(tert-Butoxy)-2-hydroxypropanoic Acid

  • MDL number MFCD28291853
  • Cas Number : 140371-02-4
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-3-(tert-butoxy)-2-hydroxypropanoic acid
Inchi Code 1S/C7H14O4/c1-7(2,3)11-4-5(8)6(9)10/h5,8H,4H2,1-3H3,(H,9,10)/t5-/m0/s1
Inchi Key LEMIPPITFFHKFG-YFKPBYRVSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid, or colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY266745&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY266745.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart