AC5H93DE8399 Accela ChemBio Inc.

5,6,7,8-Tetrahydro-2,7-naphthyridin-3(2H)-one

  • MDL number MFCD28393154
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,6,7,8-tetrahydro[2,7]naphthyridin-3-ol
Inchi Code 1S/C8H10N2O/c11-8-3-6-1-2-9-4-7(6)5-10-8/h3,5,9H,1-2,4H2,(H,10,11)
Inchi Key LHHKETSCNHQSTA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY267197&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY267197.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart