AC5H93DE839A Accela ChemBio Inc.

3-Fluoro-4-(p-tolyloxy)aniline

  • MDL number MFCD08687859
  • Cas Number : 83660-65-5
  • Molecular Weight : 217.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-(4-methylphenoxy)aniline
Inchi Code 1S/C13H12FNO/c1-9-2-5-11(6-3-9)16-13-7-4-10(15)8-12(13)14/h2-8H,15H2,1H3
Inchi Key DXBLZFBMEUKRDS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY267242&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY267242.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart