AC5H93DE839B Accela ChemBio Inc.

2-Cyclopropyl-4-ethynylthiazole

  • MDL number MFCD30071006
  • Cas Number : 1849223-21-7
  • Molecular Weight : 149.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyclopropyl-4-ethynylthiazole
Inchi Code 1S/C8H7NS/c1-2-7-5-10-8(9-7)6-3-4-6/h1,5-6H,3-4H2
Inchi Key OGKIJFKBBATRRA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY267396&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY267396.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart