AC5H93DE839F Accela ChemBio Inc.

7-Chloro-1,6-naphthyridin-2(1H)-one

  • MDL number MFCD22548048
  • Cas Number : 1345091-18-0
  • Molecular Weight : 180.5911

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Yellow to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY267403&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY267403.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart