AC5H93DE83A0 Accela ChemBio Inc.

2-Chloro-6-(trifluoromethoxy)isonicotinic Acid

  • MDL number MFCD22571780
  • Cas Number : 1221171-97-6
  • Molecular Weight : 241.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-6-(trifluoromethoxy)isonicotinic acid
Inchi Code 1S/C7H3ClF3NO3/c8-4-1-3(6(13)14)2-5(12-4)15-7(9,10)11/h1-2H,(H,13,14)
Inchi Key UCKPXDJIXHZKPQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY267487&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY267487.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart