AC5H93DE83A1 Accela ChemBio Inc.

2,3-Di-tert-butoxy-4-hydroxy-2-cyclobutenone

  • MDL number MFCD31560459
  • Cas Number : 144791-02-6
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-di-tert-butoxy-4-hydroxycyclobut-2-en-1-one
Inchi Code 1S/C12H20O4/c1-11(2,3)15-9-7(13)8(14)10(9)16-12(4,5)6/h7,13H,1-6H3
Inchi Key BLYKNWIJVHOPRO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY267489&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY267489.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart