AC5H93DE83AF Accela ChemBio Inc.

N-(4-Hydroxyphenyl)aminosulfonamide

  • MDL number MFCD25978657
  • Cas Number : 110799-26-3
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-hydroxyphenyl)sulfamide
Inchi Code 1S/C6H8N2O3S/c7-12(10,11)8-5-1-3-6(9)4-2-5/h1-4,8-9H,(H2,7,10,11)
Inchi Key WJBILEYAOOGDFI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY268122&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY268122.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart