AC5H93DE83BB Accela ChemBio Inc.

2-(4-Chlorophenyl)-5-methyl-1H-imidazole

  • MDL number MFCD09908807
  • Cas Number : 35345-09-6
  • Molecular Weight : 192.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY268360&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY268360.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart