AC5H93DE83C0 Accela ChemBio Inc.

2,4-Dichlorothieno[3,2-d]pyrimidine-6-carbaldehyde

  • MDL number MFCD29110717
  • Cas Number : 1446408-10-1
  • Molecular Weight : 233.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY268514&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY268514.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart