AC5H93DE83C1 Accela ChemBio Inc.

Methyl 2-Amino-2-(2,3-dimethylphenyl)acetate

  • MDL number MFCD16040252
  • Cas Number : 1251334-14-1
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl amino(2,3-dimethylphenyl)acetate
Inchi Code 1S/C11H15NO2/c1-7-5-4-6-9(8(7)2)10(12)11(13)14-3/h4-6,10H,12H2,1-3H3
Inchi Key QGIIYHOQEBVLPV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY268625&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY268625.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart