AC5H93DE83D1 Accela ChemBio Inc.

5-Bromo-1H-isochromen-1-one

  • MDL number MFCD26127317
  • Cas Number : 245677-36-5
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1H-isochromen-1-one
Inchi Code 1S/C9H5BrO2/c10-8-3-1-2-7-6(8)4-5-12-9(7)11/h1-5H
Inchi Key WHYKRNIHXWPVRG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY269657&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY269657.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart