AC5H93DE83DF Accela ChemBio Inc.

3,6-Dimethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD03789031
  • Cas Number : 19674-60-3
  • Molecular Weight : 140.13992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY270863&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY270863.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart