AC5H93DE83E1 Accela ChemBio Inc.

1-(2-Chloro-4-pyrimidinyl)ethanol

  • MDL number MFCD26403247
  • Cas Number : 1312535-79-7
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloropyrimidin-4-yl)ethan-1-ol
Inchi Code 1S/C6H7ClN2O/c1-4(10)5-2-3-8-6(7)9-5/h2-4,10H,1H3
Inchi Key MIUDYNZFUBDJPT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY270921&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY270921.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart