AC5H93DE83E8 Accela ChemBio Inc.

3-Bromo-2-[[(tert-butyldimethylsilyl)oxy]methyl]aniline

  • MDL number MFCD26392601
  • Cas Number : 1147531-04-1
  • Molecular Weight : 316.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2-(((tert-butyldimethylsilyl)oxy)methyl)aniline
Inchi Code 1S/C13H22BrNOSi/c1-13(2,3)17(4,5)16-9-10-11(14)7-6-8-12(10)15/h6-8H,9,15H2,1-5H3
Inchi Key HKFUSXNERADXKY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY271463&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY271463.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart