AC5H93DE83EE Accela ChemBio Inc.

Ethyl 1-Benzyl-4-chloro-6-(2-fluoro-6-methoxyphenyl)-5-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate

  • MDL number MFCD32691091
  • Molecular Weight : 429.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1-benzyl-4-chloro-6-(2-fluoro-6-methoxyphenyl)-5-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY271647&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY271647.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart