AC5H93DE83F3 Accela ChemBio Inc.

2-(Methoxycarbonyl)-1-cyclohexene-1-boronic Acid Pinacol Ester

  • MDL number MFCD22683419
  • Cas Number : 1449522-51-3
  • Molecular Weight : 266.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-1-ene-1-carboxylate
Inchi Code 1S/C14H23BO4/c1-13(2)14(3,4)19-15(18-13)11-9-7-6-8-10(11)12(16)17-5/h6-9H2,1-5H3
Inchi Key MTRUKXSEUJSVMO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY271780&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY271780.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart