AC5H93DE83F8 Accela ChemBio Inc.

N1-Boc-5-nitro-1,2-benzenediamine

  • MDL number MFCD09835423
  • Cas Number : 1201641-95-3
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-amino-5-nitrophenylcarbamate
Inchi Code 1S/C11H15N3O4/c1-11(2,3)18-10(15)13-9-6-7(14(16)17)4-5-8(9)12/h4-6H,12H2,1-3H3,(H,13,15)
Inchi Key FEDAACZCXXCKQP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY272058&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY272058.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart