AC5H93DE83FE Accela ChemBio Inc.

2-Oxo-2-(2-pyridyl)ethyl Acetate

  • MDL number MFCD19690418
  • Cas Number : 103441-80-1
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-oxo-2-(2-pyridinyl)ethyl acetate
Inchi Code 1S/C9H9NO3/c1-7(11)13-6-9(12)8-4-2-3-5-10-8/h2-5H,6H2,1H3
Inchi Key PKCHJOYORTYKQW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY272145&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY272145.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart