ADV371425085 AChemBlock

3-Methylsulfonyl-azetidine hydrochloride

  • MDL number MFCD22416367
  • Cas Number : 1400764-60-4
  • Molecular Weight : 171.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methylsulfonyl-azetidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2456
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart