ADV371425086 AChemBlock

8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

  • MDL number MFCD00151640
  • Cas Number : 28957-72-4
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Physical Form yellow solid/yellow liquid
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/8-benzyl-8-azabicyclo-3-2-1-octan-3-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2460
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart