ADV371425089 AChemBlock

exo-3-(Boc-amino)-8-azabicyclo[3.2.1]octane

  • MDL number MFCD16661075
  • Cas Number : 132234-68-5
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-3-boc-amino-8-azabicyclo-3-2-1-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3200
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart