ADV371425091 AChemBlock

exo-8-Methyl-3-amino-azabicyclo[3.2.1]octane

  • MDL number MFCD00673132
  • Cas Number : 81487-04-9
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form colorless liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/exo-8-methyl-3-amino-azabicyclo-3-2-1-octane.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+312;P305+351+338;P310
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3583
pictograms GHS05;GHS07
hstatements H302;H318
Availability details for
Quantity
  • Availability
  • Add to Cart