ADV371425092 AChemBlock

5-Bromo-2-methyl-4-nitropyridine-N-oxide

  • MDL number MFCD11977426
  • Cas Number : 62516-08-9
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-methyl-4-nitropyridine-n-oxide.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P273;P280;P301+P310;P302+352;P304+340;P305+351+338;P322;P337+313;P361;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3712
pictograms GHS06
hstatements H301;H311;H331;H315;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart