ADV371425102 AChemBlock

Pyrimidine-2-carbaldehyde

  • MDL number MFCD07778416
  • Cas Number : 27427-92-5
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/light-yellow solid
Shipping Temperature Store at -20
Country of Origin China
Product Link https://www.achemblock.com/pyrimidine-2-carbaldehyde.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338;H402+404
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2221
pictograms GHS05; GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart