ADV371425104 AChemBlock

3-Pyridazinecarbaldehyde

  • MDL number MFCD04972627
  • Cas Number : 60170-83-4
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-pyridazinecarbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2223
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart